missing translation for 'onlineSavingsMsg'
Learn More

Nonyl 4-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. H0217500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0217500G 500 g
H021725G 25 g
H0217100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0217500G Supplier TCI America Supplier No. H0217500G

Chemical Identifiers

CAS 38713-56-3
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.365
MDL Number MFCD00016483
InChI Key MBNSKHJDYXPONL-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid Nonyl Ester, Nonylparaben
PubChem CID 586708
IUPAC Name nonyl 4-hydroxybenzoate
SMILES CCCCCCCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 44°C
Color White
Quantity 500 g
Formula Weight 264.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nonyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
TSCA No