Learn More
Invitrogen™ RNase-free Microfuge Tubes
These microcentrifuge tubes are guaranteed RNase- and DNase-free.
Supplier:  Invitrogen™ AM12425
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. AM12425
Il en reste null
Description
Description
Invitrogen microcentrifuge tubes are guaranteed RNase- and DNase-free.
Features include:
  • Marked with graduations for convenience
  • Flat lids for easy labeling
  • Positive seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe
Spécifications
Identifiants chimiques
Clear
Store at room temperature.
Room Temperature
Autoclavable
Yes
Polypropylene
Invitrogen
RNase-Free, DNase-Free
Snap Cap
Microfuge Tube (RNase-free)
Spécifications
Clear
Polypropylene
Store at room temperature.
Room Temperature
Autoclavable
Yes
Microfuge Tube (RNase-free)
2 mL
500 Tubes/Pk.
Invitrogen
RNase-Free, DNase-Free
Snap Cap
Yes
Graduated
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.