missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonanoic Acid 98.0+%, TCI America™
$37.97 - $193.66
Chemical Identifiers
| CAS | 112-05-0 |
|---|---|
| Molecular Formula | C9H18O2 |
| Molecular Weight (g/mol) | 158.241 |
| MDL Number | MFCD00004433 |
| InChI Key | FBUKVWPVBMHYJY-UHFFFAOYSA-N |
| Synonym | pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a |
| PubChem CID | 8158 |
| ChEBI | CHEBI:29019 |
| IUPAC Name | nonanoic acid |
| SMILES | CCCCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
P095225ML
|
N/A | 25 mL |
Each for $37.97
|
|
|||||
|
P0952100ML
|
N/A | 100 mL |
Each for $87.34
|
|
|||||
|
P0952500ML
|
N/A | 500 mL |
Each for $193.66
|
|
|||||
Chemical Identifiers
| C9H18O2 | |
| MFCD00004433 | |
| pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a | |
| CHEBI:29019 | |
| CCCCCCCCC(=O)O |
Specifications
| 112-05-0 | |
| 254°C | |
| MFCD00004433 | |
| pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a | |
| CCCCCCCCC(=O)O | |
| 158.241 | |
| CHEBI:29019 | |
| ≥98.0% (T) | |
| Nonanoic Acid |
| Colorless | |
| C9H18O2 | |
| 3265 | |
| FBUKVWPVBMHYJY-UHFFFAOYSA-N | |
| nonanoic acid | |
| 8158 | |
| 158.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : RA6650000
TSCA : Yes