missing translation for 'onlineSavingsMsg'
Learn More

Nonanoic Acid 98.0+%, TCI America™

Catalog No. P095225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P095225ML 25 mL
P0952100ML 100 mL
P0952500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P095225ML Supplier TCI America Supplier No. P095225ML

Chemical Identifiers

CAS 112-05-0
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00004433
InChI Key FBUKVWPVBMHYJY-UHFFFAOYSA-N
Synonym pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
PubChem CID 8158
ChEBI CHEBI:29019
IUPAC Name nonanoic acid
SMILES CCCCCCCCC(=O)O

Specifications

Color Colorless
Boiling Point 254°C
Quantity 25 mL
UN Number 3265
Formula Weight 158.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Nonanoic Acid
EINECSNumber (2)-0608
RTECSNumber RA6650000
TSCA Yes