missing translation for 'onlineSavingsMsg'
Learn More

Nonanoic Acid 98.0+%, TCI America™

Supplier:  TCI America P0952100ML

Quantity 100 mL

 View more versions of this product

Catalog No. P0952100ML


Only null left

Description

Description

Specifications

Chemical Identifiers

112-05-0
158.241
FBUKVWPVBMHYJY-UHFFFAOYSA-N
8158
nonanoic acid
C9H18O2
MFCD00004433
pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
CHEBI:29019
CCCCCCCCC(=O)O

Specifications

Nonanoic Acid
Colorless
C9H18O2
MFCD00004433
pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
CCCCCCCCC(=O)O
158.241
CHEBI:29019
≥98.0% (T)
112-05-0
254°C
100 mL
3265
FBUKVWPVBMHYJY-UHFFFAOYSA-N
nonanoic acid
8158
158.24
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

RTECSNumber : RA6650000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions