missing translation for 'onlineSavingsMsg'
Learn More

Nonane 99.5+%, TCI America™

Catalog No. S02815ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02815ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02815ML Supplier TCI America Supplier No. S02815ML

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

Melting Point -51°C
Color Colorless
Boiling Point 150°C
Quantity 5 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Nonane [Standard Material for GC]
EINECSNumber (2)-0009
RTECSNumber RA6115000
TSCA Yes