missing translation for 'onlineSavingsMsg'
Learn More

Nonacosane, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014968
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014968 250 mg
111014967 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014968 Supplier Merck Emd Millipore Supplier No. 74156250MG
Only null left

Nonacosane,an aliphatic hydrocarbon, is important constituent of the leaf oil of Syzygium cumini tree, possessing anti-bacterial, anti-inflammatory and anti-ulcergenic properties.

Petroleum

Specifications

CAS 630-03-5
Physical Form Neat
Linear Formula CH3(CH2)27 CH3
MDL Number MFCD00009356
Melting Point 63°C to 66°C; 63°C to 66°C
Boiling Point 286°C (15 mmHg)
Type Analytical Standard
Quantity 250 mg
Formula Weight 408.79
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less