missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Non-Polar Fused Silica Tubing

Ideal for use with injection solvents such as alkanes, carbon disulfide, and ethers

$101.00 - $923.00

Specifications

For Use With Injection Solvents Such as Alkanes, Carbon Disulfide, and Ethers
Type Tubing
For Use With (Application) Gas Chromatography (GC)
Material Silica
View More Specs

Products 13
Catalog Number Mfr. No. Overall Length (Metric) Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Overall Length (Metric) Diameter (Metric) Inner Price Quantity & Availability  
111016172
View Documents Promotion Details
MilliporeSigma™ Supelco™
25740U
5 m 0.10 mm
Each for $155.00
Only null left
 
111004959
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA25741
5 m 0.20 mm
Each for $101.00
Only null left
 
111004960
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA25742
5 m 0.25 mm
Each for $119.00
Only null left
 
111004401
View Documents Promotion Details
MilliporeSigma™ Supelco™
25756
15 m 0.25 mm
Each for $254.00
Only null left
 
111003596
View Documents Promotion Details
MilliporeSigma™ Supelco™
25783
60 m 0.25 mm
Each for $923.00
Only null left
 
111005001
View Documents Promotion Details
MilliporeSigma™ Supelco™
25723
3 m 0.32 mm
Each for $120.00
Only null left
 
111004857
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA25743
5 m 0.32 mm
Each for $122.00
Only null left
 
111003825
View Documents Promotion Details
MilliporeSigma™ Supelco™
25770U
30 m 0.32 mm
Each for $568.00
Only null left
 
111004677
View Documents Promotion Details
MilliporeSigma™ Supelco™
25307
1 m 0.53 mm
Each for $128.00
Only null left
 
111004831
View Documents Promotion Details
MilliporeSigma™ Supelco™
25724
3 m 0.53 mm
Each for $114.00
Only null left
 
111004674
View Documents Promotion Details
MilliporeSigma™ Supelco™
25744
5 m 0.53 mm
Each for $143.00
Only null left
 
111004144
View Documents Promotion Details
MilliporeSigma™ Supelco™
25758
15 m 0.53 mm
Each for $534.90
Only null left
 
111003751
View Documents Promotion Details
MilliporeSigma™ Supelco™
25771
30 m 0.53 mm
Each for $721.00
Only null left
 
Description

Description

  • Fused silica tubing refers to tubing with a protective coating
  • Use as transfer lines, guard columns, or retention gaps
  • Can be used to make customized columns
  • Ideal for use with injection solvents such as alkanes, carbon disulfide, and ethers
  • Composition: Methyl
  • Deactivated
  • Non-polar
Specifications

Specifications

Injection Solvents Such as Alkanes, Carbon Disulfide, and Ethers
Tubing
Gas Chromatography (GC)
Silica
Videos
SDS
Documents

Documents