Learn More
MilliporeSigma™ Supelco™ Non-Polar Fused Silica Tubing
Ideal for use with injection solvents such as alkanes, carbon disulfide, and ethers
Supplier:  MilliporeSigma™ Supelco™ SGMA25741
 View more versions of this product
Catalog No. 111004959
Only null left
Description
Description
  • Fused silica tubing refers to tubing with a protective coating
  • Use as transfer lines, guard columns, or retention gaps
  • Can be used to make customized columns
  • Ideal for use with injection solvents such as alkanes, carbon disulfide, and ethers
  • Composition: Methyl
  • Deactivated
  • Non-polar
Specifications
Chemical Identifiers
Injection Solvents Such as Alkanes, Carbon Disulfide, and Ethers
Tubing
Gas Chromatography (GC)
Silica
Specifications
Injection Solvents Such as Alkanes, Carbon Disulfide, and Ethers
Gas Chromatography (GC)
5 m
Tubing
0.20 mm
Silica
Product Suggestions
Product Suggestions