missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA Classic

For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Classic with 4 sample areas per card.

$1394.75 - $1394.75

Specifications

Description Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Classic Cards.
Format Card
Sample Type Blood
For Use With (Equipment) Direct STR amplification
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
09-923-340
View Documents
QIAGEN
WB120205
100 Pk.
Pack of 100 Each for $1,394.75
Only null left
 
Description

Description

For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Classic with 4 sample areas per card.

  • QIAcard FTA Classic format with 4 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications

Specifications

Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Classic Cards.
Blood
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
FTA Classic Card
Card
Direct STR amplification
4
125 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'