missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA Classic

Catalog No. 09923340
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09923340 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09923340 Supplier QIAGEN Supplier No. WB120205

For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Classic with 4 sample areas per card.

  • QIAcard FTA Classic format with 4 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.

Specifications

Description Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Classic Cards.
Format Card
Sample Type Blood
For Use With (Equipment) Direct STR amplification
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
No. of Samples 4
Product Type FTA Classic Card
Quantity 100 Pk.
Sample Volume 125 mL