missing translation for 'onlineSavingsMsg'
Learn More

Nomega-(2-Acetamido-2-deoxy-beta-D-glucopyranosyl)-Nalpha-(tert-butoxycarbonyl)-L-asparagine 96.0+%, TCI America™

Catalog No. A1614100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1614100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1614100MG Supplier TCI America Supplier No. A1614100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137255-40-4
Molecular Formula C17H29N3O10
Molecular Weight (g/mol) 435.43
MDL Number MFCD06797037
InChI Key YVKMLCHQJOPFTE-WBXKCMFOSA-N
Synonym Nomega-(2-Acetamido-2-deoxy-beta-D-glucopyranosyl)-Nalpha-Boc-L-asparagine
PubChem CID 11004598
IUPAC Name (2S)-4-[[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(=O)NC1C(C(C(OC1NC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C)CO)O)O

Specifications

Melting Point 213°C
Color White
Quantity 100 mg
Formula Weight 435.43
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nomega-(2-Acetamido-2-deoxy-beta-D-glucopyranosyl)-Nalpha-(tert-butoxycarbonyl)-L-asparagine
TSCA No
Recommended Storage Freezer