missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ NMR/ Tube Washer
GREENER_CHOICE

Numéro de catalogue. 31501839
Change view
Click to view available options
Standard Taper Joint:
24/40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Standard Taper Joint
31501839 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 31501839 Fournisseur Synthware™ Code fournisseur T100024
Il en reste null

Features 24/40 inner joint and 10 mm hose connection.

NMR tube is inserted into the tube cleaner with its plastic cap placed on the closed end to provide support and a vacuum seal. The washing solvent is sequentially added through the side tube while a vacuum is applied through the hose connection.

Spécifications

For Use With (Application) Organic Synthesis
Type NMR Tube Washer
Material Borosilicate Glass
For Use With (Equipment) 5 mm NMR tube only
Quantity 1 Ea.
Standard Taper Joint 24/40