missing translation for 'onlineSavingsMsg'
Learn More

Nitroterephthalic Acid 98.0+%, TCI America™

$98.18 - $342.69

Chemical Identifiers

CAS 610-29-7
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007141
InChI Key QUMITRDILMWWBC-UHFFFAOYSA-N
Synonym nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u
PubChem CID 69122
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N02725G
View Documents
TCI America
N02725G
5 g
Each for $98.18
Only null left
 
N027225G
View Documents
TCI America
N027225G
25 g
Each for $342.69
Only null left
 
Description

Description

Specifications

Chemical Identifiers

610-29-7
211.129
QUMITRDILMWWBC-UHFFFAOYSA-N
69122
C8H5NO6
MFCD00007141
nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O

Specifications

610-29-7
C8H5NO6
5 g
QUMITRDILMWWBC-UHFFFAOYSA-N
211.129
211.13
Crystalline Powder
White-Yellow
MFCD00007141
nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O
69122
≥98.0% (GC,T)
Nitroterephthalic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents