missing translation for 'onlineSavingsMsg'
Learn More

Nitroterephthalic Acid 98.0+%, TCI America™

Catalog No. n02725g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02725G 5 g
N027225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02725G Supplier TCI America Supplier No. N02725G
Only null left

Chemical Identifiers

CAS 610-29-7
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007141
InChI Key QUMITRDILMWWBC-UHFFFAOYSA-N
Synonym nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u
PubChem CID 69122
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 211.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nitroterephthalic Acid