accessibility menu, dialog, popup

UserName

HOOOHONOO

CAS RN 610-29-7

IUPAC Name: 2-nitrobenzene-1,4-dicarboxylic acid
Synonyms: 2-nitrobenzene-1,4-dicarboxylic acid
Molecular Weight (g/mol): 211.13
Molecular Formula: C8H5NO6
InChi Key: QUMITRDILMWWBC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C(O)=O)C(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)