missing translation for 'onlineSavingsMsg'
Learn More

Nitrosobenzene 98.0+%, TCI America™

Catalog No. N03675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03675G 5 g
N036725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03675G Supplier TCI America Supplier No. N03675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 586-96-9
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00002059
InChI Key NLRKCXQQSUWLCH-UHFFFAOYSA-N
Synonym benzene, nitroso, nitroso-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j
PubChem CID 11473
ChEBI CHEBI:27986
IUPAC Name nitrosobenzene
SMILES C1=CC=C(C=C1)N=O

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 57°C
Quantity 5 g
UN Number 2811
Formula Weight 107.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Nitrosobenzene
EINECSNumber (3)-2752
RTECSNumber DA6497525
TSCA Yes
Recommended Storage Refrigerator