missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane 98.0+%, TCI America™

Catalog No. N0019100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0019100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0019100ML Supplier TCI America Supplier No. N0019100ML

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
Synonym methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
PubChem CID 6375
ChEBI CHEBI:77701
IUPAC Name nitromethane
SMILES C[N+]([O-])=O

Specifications

Melting Point -29°C
Color Colorless
Boiling Point 101°C
Quantity 100 mL
UN Number 1261
Formula Weight 61.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nitromethane [for Spectrophotometry]
EINECSNumber (2)-0191
RTECSNumber PA9800000
TSCA Yes