missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020183
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020183 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020183 Supplier Merck Emd Millipore Supplier No. 024841ML

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitromethane belongs to the class of residual solvents which are volatile organic compounds (VOCs) that are employed in manufacturing processes of pharmaceutical industries.

Specifications

CAS 75-52-5
Ignition Point 784°F
Vapor Density 2.1 (vs. air)
Density 1.127 g/mL (at 25°C)
Flash Point 36°C - closed cup
Linear Formula CH3 NO2
MDL Number MFCD00007400
Melting Point -29°C
Boiling Point 101.2°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 61.04
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less