missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane, 99+%, for analysis

Catalog No. AC213371000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213371000 100 mL Glass bottle
AC213375000 500 mL Glass bottle
AC213370025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC213371000 Supplier Thermo Scientific Chemicals Supplier No. 213371000

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
Synonym methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
PubChem CID 6375
ChEBI CHEBI:77701
IUPAC Name nitromethane
SMILES C[N+]([O-])=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -29.0°C
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 100.0°C to 102.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3NO2
Refractive Index 1.3805 to 1.3825
Quantity 100 mL
Residue after Evaporation 0.005% max.
Beilstein 01, 74
Fieser 01,739; 04,357; 09,323; 13,199
Specific Gravity 1.12
Merck Index 15, 6698
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide
Viscosity 0.61 mPa.s (25°C)
Formula Weight 61.04
Percent Purity 99+%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Nitromethane, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh
EINECSNumber 200-876-6

RUO – Research Use Only