missing translation for 'onlineSavingsMsg'
Learn More

Nitrite Nitrogen Standard, 1000 ppm N (3285 ppm NO2-), Ricca Chemical

$142.97 - $302.50

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5461-4
View Documents
Ricca Chemical Company
54614
120 mL Natural Poly Bottle
Each for $142.97
Only null left
 
5461-16
View Documents
Ricca Chemical Company
546116
500 mL Natural Poly Bottle
Each for $302.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7758-09-0
85.10
BXNHTSHTPBPRFX-UHFFFAOYSA-M
potassium nitrite
KNO2
MFCD00011408
516910
[K+].[O-]N=O

Specifications

Natural Poly Bottle
97.0,0.62
KNO2- in water
KNO2
Approximately 7
BXNHTSHTPBPRFX-UHFFFAOYSA-M
[K+].[O-]N=O
potassium nitrite
120 mL
ASTM
Nitrogen Standard (as Nitrite)
7758-09-0,7732-18-5
100.0,0.65
1 mL = 1 mg N as NO2
MFCD00011408
100°C
Multi-element
Water
85.10
516910
1000ppm N, 3285ppm NO2-
Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 month shelf life refrigerated.

SDS
missing translation for 'documents'

missing translation for 'documents'