missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Nitrogen Standard, 1000 ppm N (1216 ppm NH3), Ricca Chemical

$84.20 - $319.97

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5455-4
SDS View Documents
Ricca Chemical Company
54554
120 mL Natural Poly Bottle
Each for $84.20
Only null left
 
5455-16
SDS View Documents
Ricca Chemical Company
545516
500 mL Natural Poly Bottle
Each for $154.83
Only null left
 
545532
View Documents
Ricca Chemical Company
545532
1 L Natural Poly Bottle
Each for $299.67
Only null left
 
54551
View Documents
Ricca Chemical Company
54551
4 L Natural Poly Bottle
Each for $319.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
CHEBI:31206
N.Cl
ClH4N
MFCD00011420
25517
amine hydrochloride

Specifications

Natural Poly Bottle
98.0,0.37
APHA/ASTM/EPA for Ammonia Nitrogen
ClH4N
100°C
N.Cl
53.49
25517
1000ppm N, 1216ppm NH3
12125-02-9,7732-18-5
100.0,0.39
1mL = 1mg N
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
120 mL
CHEBI:31206
Nitrogen Standard (as Ammonia)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'