missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Nitrogen Standard, 1000 ppm N (1216 ppm NH3), Ricca Chemical
Supplier: Ricca Chemical Company 545532
Specifications
| Nitrogen Standard (as Ammonia) | |
| 12125-02-9,7732-18-5 | |
| 100.0,0.39 | |
| 1mL = 1mg N | |
| MFCD00011420 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 1 L | |
| CHEBI:31206 |
| Natural Poly Bottle | |
| 98.0,0.37 | |
| APHA/ASTM/EPA for Ammonia Nitrogen | |
| ClH4N | |
| 100°C | |
| N.Cl | |
| 53.49 | |
| 25517 | |
| 1000ppm N, 1216ppm NH3 |
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| CHEBI:31206 | |
| N.Cl |
| ClH4N | |
| MFCD00011420 | |
| 25517 | |
| amine hydrochloride |