missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Nitrogen Standard, 1000 ppm N (1216 ppm NH3), Ricca Chemical
Supplier: Ricca Chemical Company 54551
Specifications
| Nitrogen standard | |
| 12125-02-9,7732-18-5 | |
| MFCD00011420 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 4 L | |
| CHEBI:31206 |
| Natural Poly Bottle | |
| ClH4N | |
| 100°C | |
| N.Cl | |
| 53.49 | |
| 25517 |
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| CHEBI:31206 | |
| N.Cl |
| ClH4N | |
| MFCD00011420 | |
| 25517 | |
| amine hydrochloride |