missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurfural Diacetate, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022885
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022885 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022885 Supplier Merck Emd Millipore Supplier No. BP263

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 92-55-7
Molecular Formula C9H9NO7
Linear Formula C9H9NO7
MDL Number MFCD00003244
Quantity 50 mg
Synonym 5-Nitro-2-furaldehyde diacetate; 5-Nitrofurfurylidene diacetate
Formula Weight 243.17
Grade BP