missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurazone 98.0+%, TCI America™

Catalog No. N020025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N020025G 25 g
N0200250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N020025G Supplier TCI America Supplier No. N020025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59-87-0
Molecular Formula C6H6N4O4
Molecular Weight (g/mol) 198.14
MDL Number MFCD00003225
InChI Key IAIWVQXQOWNYOU-FPYGCLRLSA-N
Synonym nitrofurazone, nitrofural, furacilin, furacin, furacillin, aldomycin, furacine, furaldon, nifuzon, actin-n
PubChem CID 5447130
IUPAC Name [(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea
SMILES NC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O

Specifications

Melting Point 244°C
Color Yellow
pH 6.0 to 6.5 (saturated H2O)
Quantity 25 g
Formula Weight 198.14
Percent Purity ≥98.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Nitrofurazone
RTECSNumber LT7700000
TSCA Yes