missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrocyclohexane 95.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -34°C |
| Color | Yellow |
| Boiling Point | 113°C |
| Quantity | 5 g |
| UN Number | 2810 |
| Formula Weight | 129.16 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Nitrocyclohexane |
Sécurité et manipulation
| RTECSNumber | GV6600000 |
| TSCA | Yes |