missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, ACS Reagent, Reagents

Catalog No. 01337881
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337881 1 L
01337880 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337881 Supplier Labchem Inc Supplier No. C12575001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitrobenzene, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym Nitrobenzol, Oil of Mirbane
IUPAC Name nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1

Specifications

Melting Point 5.6°C to 5.8°C
Color Pale Yellow
Boiling Point 211°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.2037
Grade ACS
Physical Form Liquid
Chemical Name or Material Nitrobenzene, ACS Reagent