missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 99+%, ACS reagent

Catalog No. AC415780250
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415780250 25 mL Glass bottle
AC415785000 500 mL Glass bottle
AC415780025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC415780250 Supplier Thermo Scientific Chemicals Supplier No. 415780250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-95-3 | C6H5NO2 | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00007043
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
PubChem CID 7416
ChEBI CHEBI:27798
IUPAC Name nitrobenzene
SMILES C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Yellow
Density 1.2050g/mL
Boiling Point 210.0°C to 211.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5503 to 1.5523
Linear Formula C6H5NO2
Quantity 25 mL
Residue after Evaporation 0.005% max.
Beilstein 05, 233
Fieser 02,295; 07,251; 08,358; 17,16
Specific Gravity 1.205
Merck Index 15, 6674
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,benzene,ether and oils
Formula Weight 123.11
Percent Purity 99+%
Grade ACS Reagent
Titratable Acid 0.0005 meq/g (Water soluble) max
Physical Form Liquid
Chemical Name or Material Nitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.
Toxic if sw
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
EINECSNumber 202-716-0
RTECSNumber DA6475000
TSCA TSCA

RUO – Research Use Only