Learn More
Nitro Blue Tetrazolium chloride, 98+%
Description
Intense blue dye used in histochemistry determinations.Nitro Blue Tetrazolium chloride is used to confirm diagnosis of chronic granulomatous disease. It is a microscopic stain, which is used to detect dehydrogenases and alkaline phosphatase. It acts as an NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase. Further, it is used in immunology for detection of alkaline phosphatase. In addition to this, it acts as an oxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Intense blue dye used in histochemistry determinations.Nitro Blue Tetrazolium chloride is used to confirm diagnosis of chronic granulomatous disease. It is a microscopic stain, which is used to detect dehydrogenases and alkaline phosphatase. It acts as an NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase. Further, it is used in immunology for detection of alkaline phosphatase. In addition to this, it acts as an oxidant.
Solubility
Soluble in water, ethanol and methanol.
Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼190°C (decomposition) |
| Sensitivity | Light sensitive |
| Beilstein | 4115923 |
| Color | Yellow |
| Solubility Information | Soluble in water,ethanol and methanol. |
| Quantity | 5 g |
| Physical Form | Crystalline Powder |
| Formula Weight | 817.65 |
| Percent Purity | 99% |
| Chemical Name or Material | Nitro Blue Tetrazolium chloride |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 206-067-4 |
| RTECSNumber | XF8045000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only