missing translation for 'onlineSavingsMsg'
Learn More

Nitrite Nitrogen Standard, 1000 ppm N (3285 ppm NO2-), Ricca Chemical

Catalog No. R5444900120
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5444900120 120 mL Amber Glass Bottle
R5444900500 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R5444900120 Supplier Ricca Chemical Company Supplier No. R5444900120C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-09-0 , 7732-18-5
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 1000ppm N as Nitrite (3285ppm NO2-)
CAS Min % 97.0, 0.62
CAS Max % 100.0, 0.65
Equivalence 1 mL = 1 mg N
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Grade ACS
Chemical Name or Material Nitrite Standard
Show More Show Less