missing translation for 'onlineSavingsMsg'
Learn More

Nitrite ion standard Solution, 0.1M NO2-, For ion-selective electrodes, MilliporeSigma™ Supelco™

Catalog No. 111015458
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015458 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015458 Supplier Merck Emd Millipore Supplier No. 72586100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

CAS 7632-00-0
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
Synonym Sodium nitrite solution

Specifications

Physical Form Liquid
Quantity 100 mL
Linear Formula NaNO2
Formula Weight 69.00
Concentration or Composition (by Analyte or Components) 0.1 M NO 2 -
Density 1.00 g/mL (at 20°C)
Chemical Name or Material Sodium Nitrite
ShelfLife Limited shelf life, expiry date on the label