missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrite (as Nitrogen) Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N, 1mL = 3.28mg NO2- ), LabChem™
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS Max % | 99.39, 0.61 |
| Identification | Passes Test |
| Density | 1g/mL |
| Physical Form | Liquid |
| Concentration | 1000 ppm ±10 ppm (1mL = 1mg N, 1mL = 3.28mg NO2- ) |
| Linear Formula | KNO2 |
| Color | Colorless |
| Solubility | Soluble in water |
| Quantity | 500 mL |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Avoid release to the environment. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Refrigerate |