missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotris(methylenephosphonic Acid) (ca. 50% in Water, ca. 2.2mol/L), TCI America™

Catalog No. N0474500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0474500ML 500 mL
N047425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0474500ML Supplier TCI America Supplier No. N0474500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6419-19-8
Molecular Formula C3H12NO9P3
Molecular Weight (g/mol) 299.048
MDL Number MFCD00002138
InChI Key YDONNITUKPKTIG-UHFFFAOYSA-N
Synonym nitrilotris methylene triphosphonic acid, tris phosphonomethyl amine, ferrofos 509, aminotrimethylene phosphonic acid, dowell l 37, dequest 2000, nitrilotrimethylphosphonic acid, nitrilotris methylenephosphonic acid, aminotris methylphosphonic acid, aminotri methylenephosphonic acid
PubChem CID 16698
IUPAC Name [bis(phosphonomethyl)amino]methylphosphonic acid
SMILES C(N(CP(=O)(O)O)CP(=O)(O)O)P(=O)(O)O

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 299.05
Physical Form Liquid
Chemical Name or Material Nitrilotris(methylenephosphonic Acid) (ca. 50% in Water, ca. 2.2mol/L)
EINECSNumber (2)-1866&(2)-2009
RTECSNumber SZ9860000
TSCA Yes