missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic Acid, ACS Grade, ≥98%, MP Biomedicals™

Catalog No. ICN15255180
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15255180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15255180 Supplier MP Biomedicals Inc Supplier No. 0215255180

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Color White
Assay Percent Range ≥98%
Quantity 100 g
Formula Weight 191.14
Grade ACS Reagent
Physical Form Solid
Chemical Name or Material Nitrilotriacetic Acid
Recommended Storage Store at room temperature (15° to 30°C)