missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, ACS, 68-70%, Spectrum™ Chemical

$466.73 - $13087.10

Chemical Identifiers

CAS 7697-37-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612005
View Documents
Spectrum Chemical Mfg Cor
N1075500MLGL
500 mL Amber Glass Bottle
Each for $466.73
Only null left
 
18612003
View Documents
Spectrum Chemical Mfg Cor
N107525LTPV
2.5 L Poly-Coated Glass Bottle
Each for $663.10
Only null left
 
18612004
View Documents
Spectrum Chemical Mfg Cor
N1075200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7697-37-2

Specifications

Colorless
1
Amber Glass Bottle
500 mL
Reagent
7697-37-2,7732-18-5
0.2 ppm
5 ppm
68 to 70%
Nitric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.