missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, ACS, 68-70%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor N1075200LTBL

 View more versions of this product

Catalog No. 18612004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Nitric Acid
7697-37-2,7732-18-5
0.2 ppm
5 ppm
68 to 70%
Colorless
1
Drum
200 L
Reagent

Chemical Identifiers

7697-37-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.