missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, ACS, 68-70%, Spectrum™ Chemical

Catalog No. 18612005
Change view
Click to view available options
Quantity:
500 mL
2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612005 500 mL Amber Glass Bottle
18612003 2.5 L Poly-Coated Glass Bottle
18612004 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612005 Supplier Spectrum Chemical Mfg Cor Supplier No. N1075500MLGL

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5

Specifications

Appearance Colorless
CAS Max % 1
Heavy Metals 0.2 ppm
Packaging Amber Glass Bottle
Residue after Ignition 5 ppm
Quantity 500 mL
Percent Purity 68 to 70%
Grade Reagent
Chemical Name or Material Nitric Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.