missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 2.00 N, Ricca Chemical

Catalog No. 543216
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
543216 500 mL Natural Poly Bottle
543232 1 L Natural Poly Bottle
54325PT 2.5 L Safety-coated Glass Bottle
R543200020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 543216 Supplier Ricca Chemical Company Supplier No. 543216
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
Concentration 2.00Normal
CAS Min % 82.0, 15.81
CAS Max % 86.0, 16.45
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Nitric Acid