missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 10% (v/v), Ricca Chemical

Catalog No. 53205PT
Change view
Click to view available options
Quantity:
2.5 L
4 L
Packaging:
Natural Poly Bottle
Safety-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
53205PT 2.5 L Safety-coated Glass Bottle
53201 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53205PT Supplier Ricca Chemical Company Supplier No. 53205PT

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
Concentration 10% (v/v)
CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Packaging Safety-coated Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 2.5 L
Physical Form Liquid
Percent Purity 10% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid
Show More Show Less