missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 0.500 N (N/2), Ricca Chemical

Supplier:  Ricca Chemical Company R54250001A

Quantity 1 L
Packaging Natural Poly Bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. R54250001A

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

7697-37-2 , 7732-18-5
63.012
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
[N+](=O)(O)[O-]
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid

Spécifications

Nitric Acid
7697-37-2 , 7732-18-5
Natural Poly Bottle
0.5000 ± 0.0005 N at 20°C
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
0.5N
Odorless to slightly pungent
Aqueous Solution
1 L
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
[N+](=O)(O)[O-]
63.012
CHEBI:48107
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions