missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 0.0200 N (N/50), Ricca Chemical
Chemical Identifiers
| CAS | 7697-37-2 , 7732-18-5 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.012 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | [N+](=O)(O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
538516
|
N/A | 500 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
538532
|
N/A | 1 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7697-37-2 , 7732-18-5 | |
| 63.012 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| [N+](=O)(O)[O-] |
| HNO3 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
Specifications
| 7697-37-2 , 7732-18-5 | |
| 100.0, 0.2 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| [N+](=O)(O)[O-] | |
| 63.012 | |
| CHEBI:48107 |
| 98.0, 0.19 | |
| HNO3 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| Nitric Acid |