missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 0.0200 N (N/50), Ricca Chemical
Supplier: Ricca Chemical Company 538516
| Quantity | 500 mL |
|---|---|
| Packaging | Natural Poly Bottle |
Chemical Identifiers
| 7697-37-2 , 7732-18-5 | |
| 63.012 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| [N+](=O)(O)[O-] |
| HNO3 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
Specifications
| Nitric Acid | |
| 98.0, 0.19 | |
| Natural Poly Bottle | |
| HNO3 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 |