missing translation for 'onlineSavingsMsg'
Learn More

Nitrate (as Nitrogen) Standard, 1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 4.43mg NO3-), Certified, LabChem™

Catalog No. p-5388780
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC179107 125 mL Poly Bottle
LC179101 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC179107 Supplier LabChem Supplier No. LC179107
Only null left

Chemical Identifiers

CAS 7757-79-1 , 67-66-3 , 64-17-5 , 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 99.08, 0.72, 0.198, 0.002
Identification Passes Test
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg N, 1mL = 4.43mg NO3-)
Linear Formula KNO3
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 101.1
Grade Certified
Assay as N
Chemical Name or Material Nitrate (as Nitrogen) Standard, 1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 4.43mg NO3-)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause cancer (oral, inhalation).
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature