missing translation for 'onlineSavingsMsg'
Learn More

Nitarsone 98.0+%, TCI America™

Catalog No. N0964200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0964200MG 200 mg
N09641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0964200MG Supplier TCI America Supplier No. N0964200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-72-6
Molecular Formula C6H6AsNO5
Molecular Weight (g/mol) 247.04
MDL Number MFCD00024575
InChI Key FUUFQLXAIUOWML-UHFFFAOYSA-N
Synonym 4-Nitrophenylarsonic Acid, 4-Nitrobenzenearsonic Acid
PubChem CID 66826
ChEBI CHEBI:62629
IUPAC Name (4-nitrophenyl)arsonic acid
SMILES O[As](O)(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 300°C
Color Yellow
Quantity 200 mg
UN Number 3465
Formula Weight 247.04
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nitarsone