missing translation for 'onlineSavingsMsg'
Learn More

Nisoldipine 98.0+%, TCI America™

Catalog No. N09005G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09005G 5 g
N09001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09005G Supplier TCI America Supplier No. N09005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63675-72-9
Molecular Formula C20H24N2O6
Molecular Weight (g/mol) 388.42
MDL Number MFCD00478055
InChI Key VKQFCGNPDRICFG-UHFFFAOYNA-N
Synonym 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Isobutyl Methyl Ester, Isobutyl Methyl 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate
PubChem CID 4499
ChEBI CHEBI:76917
IUPAC Name 3-methyl 5-(2-methylpropyl) 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OCC(C)C

Specifications

Melting Point 153°C
Color Yellow
Quantity 5 g
Formula Weight 388.42
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Nisoldipine
RTECSNumber US7975600
TSCA No