Learn More
Abcam Inc NIR-797-isothiocyanate, Near-infrared fluorescent dye, 25MG
Numéro de catalogue. 502705339
Encompass
MW 880.2 Da, Purity >90%. Near-infrared fluorescent dye. Cyanine dye for labeling protein. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502705339 Fournisseur Abcam Inc Code fournisseur AB14528025MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 880.2 Da, Purity >90%. Near-infrared fluorescent dye. Cyanine dye for labeling protein. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement