missing translation for 'onlineSavingsMsg'
Learn More

Nioxime 98.0+%, TCI America™

Catalog No. C04835G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04835G 5 g
C04831G 1 g
C048325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C04835G Supplier TCI America Supplier No. C04835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 492-99-9
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00001663
InChI Key CUNNCKOPAWXYDX-KQQUZDAGSA-N
Synonym 1,2-cyclohexanedione dioxime, nioxime, 1z,2z-cyclohexane-1,2-dione dioxime, nioxim, 1,2-cyclohexanedione, dioxime, 1,2-bis hydroxyimino cyclohexane, unii-g1qt7su5a8, cyclohexane-1,2-dione dioxime, 1,2-cyclohexanedione, 1,2-dioxime, g1qt7su5a8
PubChem CID 10300
IUPAC Name N-[(1E,2E)-2-(hydroxyimino)cyclohexylidene]hydroxylamine
SMILES O\N=C1/CCCC/C/1=N\O

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 5 g
Formula Weight 142.16
Percent Purity ≥98.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Nioxime