missing translation for 'onlineSavingsMsg'
Learn More

Niobium gauze, 40 mesh woven from 0.178mm (0.007 in.) dia. wire, Thermo Scientific Chemicals

Catalog No. AA40916FL
Change view
Click to view available options
Quantity:
75 x 75 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40916FL 75 x 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40916FL Supplier Thermo Scientific Chemicals Supplier No. 040916FL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Filtration, electrodes

Filtration, electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Quantity 75 x 75 mm
Mesh Size 40 Mesh
Form Wire Cloth
Chemical Name or Material Niobium gauze, Woven from 0.178mm (0.007 in.) dia. wire
EINECSNumber 231-113-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only