missing translation for 'onlineSavingsMsg'
Learn More

Nimodipine, Thermo Scientific™

Catalog No. p-7077237
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC329280010 1g
AC329280050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC329280010 Supplier Thermo Scientific Chemicals Supplier No. 329280010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 66085-59-4
Molecular Formula C21H26N2O7
Molecular Weight (g/mol) 418.45
MDL Number MFCD00153848
InChI Key UIAGMCDKSXEBJQ-UHFFFAOYNA-N
Synonym nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
PubChem CID 4497
ChEBI CHEBI:7575
IUPAC Name 3-O-(2-methoxyethyl) 5-O-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C

Specifications

Loss on Drying 0.5% max. (105°C, 4 h)
Infrared Spectrum Authentic
Quantity 1g
Merck Index 15,6636
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso
Formula Weight 418.44
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Nimodipine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical attention/advice if you feel unwell.
EINECSNumber 266-127-

RUO – Research Use Only